HPLC Analysis of 3,5-DNB-Tryptophan Methyl Ester Enantiomers on Astec® CYCLOBOND I 2000 SN
![HPLC Analysis of 3,5-DNB-Tryptophan Methyl Ester Enantiomers on Astec® CYCLOBOND I 2000 SN application for HPLC](/deepweb/assets/sigmaaldrich/product/images/416/004/fc1023e6-180f-406e-b4fc-6907b89ec21a/800/fc1023e6-180f-406e-b4fc-6907b89ec21a.jpg)
Materials
standard
Product No.
Description
Pricing
CONDITIONS
column
CYCLOBOND I 2000 SN, 25 cm x 4.6 mm I.D., 5 μm particles (20524AST)
mobile phase
ethanol
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null