Skip to Content
Merck
All Photos(2)

Key Documents

348562

Sigma-Aldrich

(S)-(+)-2-Methylbutyric anhydride

95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$1,486.00

R$1,486.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$1,486.00

About This Item

Linear Formula:
[C2H5CH(CH3)CO]2O
CAS Number:
Molecular Weight:
186.25
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

R$1,486.00


Check Cart for Availability

Request a Bulk Order

Assay

95%

form

liquid

optical activity

[α]20/D +35°, neat

refractive index

n20/D 1.418 (lit.)

bp

60 °C/0.2 mmHg (lit.)

density

0.934 g/mL at 25 °C (lit.)

functional group

anhydride
ester

SMILES string

CC[C@H](C)C(=O)OC(=O)[C@@H](C)CC

InChI

1S/C10H18O3/c1-5-7(3)9(11)13-10(12)8(4)6-2/h7-8H,5-6H2,1-4H3/t7-,8-/m0/s1

InChI key

WRTPVONNQPWNRH-YUMQZZPRSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian Schwindl et al.
Phytochemistry, 144, 58-70 (2017-09-11)
A methanolic extract of Juglans regia L. leaves was fractioned by various chromatographic techniques yielding a total of 40 metabolites belonging to megastigmane, tetralone, phenylpropanoid, neolignane and juglone glycosides. Ten unknown megastigmane glucoside derivatives (juglanionosides A-K, 1-10) and six unknown

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service