Skip to Content
Merck
All Photos(3)

Documents

245526

Sigma-Aldrich

(S)-(+)-2-Methylbutyric acid

98%

Synonym(s):

(2S)-2-methylbutanoic acid, (S)-2-methylbutanoate, (S)-2-methylbutanoic acid, (S)-2-methylbutyric acid, 2-methylbutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CO2H
CAS Number:
Molecular Weight:
102.13
Beilstein:
1720485
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

optical activity

[α]20/D +19°, neat

optical purity

ee: 98% (GLC)

refractive index

n20/D 1.406 (lit.)

bp

78 °C/15 mmHg (lit.)

density

0.938 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC[C@H](C)C(O)=O

InChI

1S/C5H10O2/c1-3-4(2)5(6)7/h4H,3H2,1-2H3,(H,6,7)/t4-/m0/s1

InChI key

WLAMNBDJUVNPJU-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(+)-2-Methylbutyric acid may be used in the preparation of the stereoisomers of (R)-lavandulyl (S)-2-methylbutanoate and (R)-maconelliyl (S)-2-methylbutanoate, which are female sex pheromones found in pink hibiscus mealybug.
Chiral starting material

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sex pheromone of the pink hibiscus mealybug, Maconellicoccus hirsutus, contains an unusual cyclobutanoid monoterpene.
Zhang A, et al.
Proceedings of the National Academy of Sciences of the USA, 101(26), 9601-9606 (2004)
Lauren E Kelley et al.
Journal of agricultural and food chemistry, 66(10), 2414-2421 (2017-01-07)
Potent odorants in Native spearmint, Scotch spearmint, and Macho mint oils were determined by the combined use of gas chromatography-olfactometry (GCO), gas chromatography-mass spectrometry (GC-MS), and aroma extract dilution analysis (AEDA). Of the 85 odorants detected, ( R)-(-)-carvone was the
Gregory Kaler et al.
The Journal of biological chemistry, 282(33), 23841-23853 (2007-06-08)
Organic anion transporters (OATs, SLC22) interact with a remarkably diverse array of endogenous and exogenous organic anions. However, little is known about the structural features that determine their substrate selectivity. We examined the substrate binding preferences and transport function of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service