Skip to Content
Merck
All Photos(1)

Key Documents

285978

Sigma-Aldrich

D-Glucose 6-phosphate sodium salt

Synonym(s):

D(+)-Glucopyranose 6-phosphate sodium salt, G-6-P Na, Robison ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12NaO9P
CAS Number:
Molecular Weight:
282.12
Beilstein:
5787568
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

optical activity

[α]25/D +33°, c = 10 in H2O

mp

204 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

[Na+].O[C@@H]1O[C@H](COP(O)([O-])=O)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C6H13O9P.Na/c7-3-2(1-14-16(11,12)13)15-6(10)5(9)4(3)8;/h2-10H,1H2,(H2,11,12,13);/q;+1/p-1/t2-,3-,4+,5-,6-;/m1./s1

InChI key

ZALKNDISPIVVKC-WYRLRVFGSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pascal Belin et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(18), 7426-7431 (2009-05-07)
The gene encoding the cytochrome P450 CYP121 is essential for Mycobacterium tuberculosis. However, the CYP121 catalytic activity remains unknown. Here, we show that the cyclodipeptide cyclo(l-Tyr-l-Tyr) (cYY) binds to CYP121, and is efficiently converted into a single major product in
Hwayoun Lee et al.
Journal of toxicology and environmental health. Part A, 77(22-24), 1409-1418 (2014-10-25)
Cytochrome P450 2A6 (P450 2A6) is the major enzyme responsible for the oxidation of coumarin, nicotine, and tobacco-specific nitrosamines in human liver. In this study, the catalytic turnover of coumarin oxidation was improved by directed-evolution analysis of P450 2A6 enzyme.
Qi Wang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 983-984, 39-46 (2015-01-24)
Coumarins are an important group of bioactive constituents in licorice (Glycyrrhiza uralensis), a worldwide popular herbal medicine. This study aims to elucidate the metabolism of two major licorice coumarins, glycyrin and glycyrol in rats. After oral administration of 40mg/kg glycyrin
Massimiliano Paganelli et al.
Stem cells and development, 23(12), 1377-1391 (2014-02-20)
Understanding the mechanisms triggering hepatogenic differentiation of stem/progenitor cells would be useful for studying postnatal liver regeneration and development of liver cell therapies. Many evidences support the involvement of Sox9 transcription factor in liver development. Here, we investigate the possibility

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service