Skip to Content
Merck
All Photos(3)

Documents

G6526

Sigma-Aldrich

D-Glucose 6-phosphate potassium salt

≥95% anhydrous basis (enzymatic)

Synonym(s):

D(+)-Glucopyranose 6-phosphate potassium salt, Robison ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H12O9PK
CAS Number:
Molecular Weight:
298.23
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥95% anhydrous basis (enzymatic)

form

powder

color

white

solubility

water: 50 mg/mL, clear to hazy, colorless

cation traces

K: 10.16-15.88% (ICP anhydrous)

storage temp.

−20°C

SMILES string

[K].OC1OC(COP(O)(O)=O)C(O)C(O)C1O

InChI

1S/C6H13O9P.K.H/c7-3-2(1-14-16(11,12)13)15-6(10)5(9)4(3)8;;/h2-10H,1H2,(H2,11,12,13);;

InChI key

UTZCQRPSJJQUEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Treatment of rat pancreatic islets with the glucose metabolite D-glucose 6-phosphate elevates the level of ATP-dependent Ca2+ within the endoplasmic reticulum indicating it is a key regulator of Ca2+ storage in the islets.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B A Wolf et al.
The Journal of biological chemistry, 261(35), 16284-16287 (1986-12-15)
Glucose stimulation of islets is coupled with the rapid intracellular release of myo-inositol 1,4,5-trisphosphate (IP3) and arachidonic acid which in turn mobilize Ca2+ stored in the endoplasmic reticulum (ER). The metabolism of glucose is required for insulin secretion although the

Articles

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service