Skip to Content
Merck
All Photos(1)

Key Documents

265292

Sigma-Aldrich

2,4-Difluorobenzoyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3COCl
CAS Number:
Molecular Weight:
176.55
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.5157 (lit.)

density

1.437 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(c(F)c1)C(Cl)=O

InChI

1S/C7H3ClF2O/c8-7(11)5-2-1-4(9)3-6(5)10/h1-3H

InChI key

JSWRVDNTKPAJLB-UHFFFAOYSA-N

Application

2,4-Difluorobenzoyl chloride was used in the synthesis of 3-oxocyclohex-1-enyl 2,4-difluorobenzoate[1].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and conjugate additions of 2, 3, 4, 9-tetrahydro-1H-xanthene-1, 9-diones.
Gabbutt CD, et al.
Journal of the Chemical Society. Perkin Transactions 1, 12, 1819-1824 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service