Skip to Content
Merck
All Photos(1)

Key Documents

924520

Sigma-Aldrich

4-Methoxybenzyl chloride

Synonym(s):

4-(Chloromethyl)anisole

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
R$1,391.00

R$1,391.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
R$1,391.00

About This Item

Linear Formula:
CH3OC6H4CH2Cl
CAS Number:
Molecular Weight:
156.61
Beilstein:
606667
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.22

R$1,391.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

refractive index

n20/D 1.548 (lit.)

bp

117-118 °C/14 mmHg (lit.)

mp

−1 °C (lit.)

density

1.155 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

COc1ccc(CCl)cc1

InChI

1S/C8H9ClO/c1-10-8-4-2-7(6-9)3-5-8/h2-5H,6H2,1H3

InChI key

MOHYOXXOKFQHDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methoxybenzyl chloride can be used as a reactant:
  • To synthesize 4-methoxybenzyl esters by reacting with cyclic anhydrides.[1]
  • In the mono-alkylation of trans-1,2-diaminocyclohexane.[2]
It can also be used as a:
  • Protecting group for amino acids during peptide synthesis using triethylamine as a base.[1]
  • Co-catalyst in the intramolecular carboamination of aminodienes to N-heterocyclesvia benzyl-group transfer.[3]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service