Skip to Content
Merck
All Photos(1)

Key Documents

15513

Sigma-Aldrich

N-Boc-N′-Fmoc-diaminoacetic acid

technical, ≥95.0% (HPLC)

Synonym(s):

2-(9H-Fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid, 2-[[(1,1-Dimethylethoxy)carbonyl]amino]-2-[[(9H-fluoren-9-ylmethoxy)carbonyl]amino]acetic acid, Parexan, Piresan, Pyganic, Pyrellin, Pyrethrine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O6
CAS Number:
Molecular Weight:
412.44
Beilstein:
6163483
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

grade

technical

Assay

≥95.0% (HPLC)

form

liquid

application(s)

peptide synthesis

functional group

Boc
Fmoc

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)NC(NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C22H24N2O6/c1-22(2,3)30-21(28)24-18(19(25)26)23-20(27)29-12-17-15-10-6-4-8-13(15)14-9-5-7-11-16(14)17/h4-11,17-18H,12H2,1-3H3,(H,23,27)(H,24,28)(H,25,26)

InChI key

AWCFKRJSLGKRNA-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service