Skip to Content
Merck
All Photos(1)

Key Documents

86971

Sigma-Aldrich

Fmoc-Dab(Boc)-OH

≥97.0% (HPLC)

Synonym(s):

Nα-Fmoc-Nγ-Boc-L-2,4-diaminobutyric acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$894.00

R$894.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$894.00

About This Item

Empirical Formula (Hill Notation):
C24H28N2O6
CAS Number:
Molecular Weight:
440.49
Beilstein:
6250328
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

R$894.00


Check Cart for Availability

Request a Bulk Order

Assay

≥97.0% (HPLC)

form

powder

optical activity

[α]20/546 −14.5±1°, c = 1% in methanol

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Boc
Fmoc

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)NCC[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C24H28N2O6/c1-24(2,3)32-22(29)25-13-12-20(21(27)28)26-23(30)31-14-19-17-10-6-4-8-15(17)16-9-5-7-11-18(16)19/h4-11,19-20H,12-14H2,1-3H3,(H,25,29)(H,26,30)(H,27,28)/t20-/m0/s1

InChI key

LIWKOFAHRLBNMG-FQEVSTJZSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronald Domalaon et al.
Amino acids, 46(11), 2517-2530 (2014-07-30)
Taking a minimalistic approach in efforts to lower the cost for the development of new synthetic antimicrobial peptides, ultrashort cationic lipopeptides were designed to mimic the amphiphilic nature crucial for their activity but with only a very short peptide sequence
Tony Velkov et al.
Cell chemical biology, 25(4), 380-391 (2018-02-06)
Resistance to the last-resort antibiotic colistin is now widespread and new therapeutics are urgently required. We report the first in toto chemical synthesis and pre-clinical evaluation of octapeptins, a class of lipopeptides structurally related to colistin. The octapeptin biosynthetic cluster consisted
Kuo-Yuan Hung et al.
European journal of medicinal chemistry, 136, 154-164 (2017-05-12)
The 20-residue linear peptide A20FMDV2 has been shown to exhibit high selectivity and affinity for the tumour-related αvβ6 integrin and has potential as a vector for therapeutic drugs. However, it exhibits poor half-life in plasma in part due to its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service