Skip to Content
Merck
All Photos(1)

Key Documents

133469

Sigma-Aldrich

3-Amino-5,6-dimethyl-1,2,4-triazine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8N4
CAS Number:
Molecular Weight:
124.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

210-212 °C (lit.)

SMILES string

Cc1nnc(N)nc1C

InChI

1S/C5H8N4/c1-3-4(2)8-9-5(6)7-3/h1-2H3,(H2,6,7,9)

InChI key

UIKGLXJNZXSPGV-UHFFFAOYSA-N

General description

3-Amino-5,6-dimethyl-1,2,4-triazine reacts with silver trifluoro­methane­sulfonate to yield tris(3-amino-5,6-dimethyl-1,2,4-triazine-κN)silver(I) trifluromethane-sulfonate-3-amino-5,6-dimethyl-1,2,4-triazine. It reacts with an excess of benzoyl chloride in the presence of triethylamine in refluxing chloroform to give 3-dibenzoylamino-5,6-dimethyl-1,2,4-triazine.

Application

3-Amino-5,6-dimethyl-1,2,4-triazine was used in intramolecular cyclization reaction of various 1,2-bis(amidinohydrazone)s.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction of 3-amino-5, 6-dimethyl-1, 2, 4-triazine with electrophiles.
Suzuki T, et al.
Journal of Heterocyclic Chemistry, 23(3), 935-939 (1986)
A novel intramolecular cyclization reaction: Facile solvent-free microscale synthesis suitable for rapid production of libraries of 3-amino-1, 2, 4-triazines and for combinatorial chemistry.
Matikainen JKT and Elo HA.
Microchimica Acta, 146(1), 49-53 (2004)
Yu-Hang Jiang et al.
Acta crystallographica. Section E, Structure reports online, 67(Pt 11), m1526-m1527 (2012-01-06)
The asymmetric unit of the title compound, [Ag(C(5)H(8)N(4))(3)](CF(3)O(3)S)·C(5)H(8)N(4), contains two cations, two anions and two uncoordinated 3-amino-5,6-dimethyl-1,2,4-triazine (admt) ligands. It was prepared from the reaction of silver trifluoro-methane-sulfonate and admt in a 2:3 molar ratio. Both silver(I) ions are bonded

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service