Skip to Content
Merck
All Photos(2)

Key Documents

308153

Sigma-Aldrich

N-Propyl-1,3-propanediamine

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$657.00

R$657.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
R$657.00

About This Item

Linear Formula:
CH3CH2CH2NH(CH2)3NH2
CAS Number:
Molecular Weight:
116.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$657.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.4451 (lit.)

bp

169 °C (lit.)

density

0.841 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCCNCCCN

InChI

1S/C6H16N2/c1-2-5-8-6-3-4-7/h8H,2-7H2,1H3

InChI key

OWKYZAGJTTTXOK-UHFFFAOYSA-N

Application

N-Propyl-1,3-propanediamine was used in the synthesis of 1-benzotriazolylmethyl-3-propylhexahydropyrimidine[1].

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed Salman et al.
Genes, 11(10) (2020-10-04)
In this study, we seek to exclude other pathophysiological mechanisms by which Frmd7 knock-down may cause Idiopathic Infantile Nystagmus (IIN) using the Frmd7.tm1a and Frmd7.tm1b murine models. We used a combination of genetic, histological and visual function techniques to characterize
Alan R Katritzky et al.
The Journal of organic chemistry, 67(9), 3115-3117 (2002-04-27)
1-Benzotriazolylmethyl-3-propylhexahydropyrimidine (1) and 1,3-bis(1H-1,2,3-benzotriazol-1-ylmethyl)-1,2,3,4-tetrahydroquinazoline (3) were readily prepared by reactions of N-propyl-1,3-propanediamine or 2-aminobenzylamine with benzotriazole and formaldehyde, respectively. Intermediate 1 reacted with alkyl and aryl Grignard reagents to produce N,N'-unsymmetrically substituted hexahydropyrimidines 2a,b in 90 and 92% yields, respectively.
C M Maragos et al.
Cancer research, 53(3), 564-568 (1993-02-01)
Cell-mediated antitumor effects have, in part, been attributed to the production of NO. Compounds which generate NO might, therefore, be useful in attenuating the growth of tumor cells. Six nitric oxide/nucleophile adducts that release NO spontaneously in solution were tested

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service