Recommended Products
Assay
≥98.5% (T)
form
solid
impurities
≤0.5% water
mp
130-135 °C
SMILES string
[Cl-].N#CCCC[n+]1ccccc1
InChI
1S/C9H11N2.ClH/c10-6-2-5-9-11-7-3-1-4-8-11;/h1,3-4,7-8H,2,5,9H2;1H/q+1;/p-1
InChI key
VOKQCUHMWFKHFE-UHFFFAOYSA-M
General description
Nitrile-functionalized ionic liquid showing superior characteristics in Suzuki and Stille coupling reactions.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The American journal of pathology, 190(3), 674-688 (2020-01-24)
miRNAs, a well-known group of noncoding RNAs, contribute to the pathogenesis of multiple diseases, including colitis-associated cancer (CAC). Our recent findings indicate that proinflammatory cytokines up-regulate c-MYC level, which subsequently activates cullin 4A and 4B (CUL4A/4B) and CRL4DCAF4 E3 ligases
Journal of the American Chemical Society, 126(48), 15876-15882 (2004-12-02)
A series of relatively low-cost ionic liquids, based on the N-butyronitrile pyridinium cation [C(3)CNpy](+), designed to improve catalyst retention, have been prepared and evaluated in Suzuki and Stille coupling reactions. Depending on the nature of the anion, these salts react
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service