Skip to Content
Merck
All Photos(2)

Key Documents

292796

Sigma-Aldrich

Benzyldimethyltetradecylammonium chloride dihydrate

98%

Synonym(s):

N,N-dimethyl-N-tetradecyl-benzenemethanaminium chloride dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
R$2,495.00

R$2,495.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
R$2,495.00

About This Item

Linear Formula:
CH3(CH2)13N(Cl)(CH3)2CH2C6H5 · 2H2O
CAS Number:
Molecular Weight:
404.07
Beilstein:
4062599
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

R$2,495.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

mp

63-65 °C (lit.)

SMILES string

[Cl-].[H]O[H].[H]O[H].CCCCCCCCCCCCCC[N+](C)(C)Cc1ccccc1

InChI

1S/C23H42N.ClH.2H2O/c1-4-5-6-7-8-9-10-11-12-13-14-18-21-24(2,3)22-23-19-16-15-17-20-23;;;/h15-17,19-20H,4-14,18,21-22H2,1-3H3;1H;2*1H2/q+1;;;/p-1

InChI key

YLPZVNXGKBIJBW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Benzyldimethyltetradecylammonium chloride dehydrate is a general cationic surfactant[1] that is also used as a phase transfer catalyst in:
  • The synthesis of triazine containing fluoropolymer with linear dielectric properties.[2]
  • The preparation of mixed micelle with tetradecyltrimethylammonium (TTAB) and tetradecyltriphenylphosphonium bromides.[3]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new fluoropolymer having triazine rings as a dielectric material: synthesis and properties.
Zhou J, et al.
Polym. Chem., 8(39), 6173-6180 (2017)
On the new possibility of applying oscillating liquidmembrane systems for molecular recognition substances responsible for taste.
Szpakowska M, et al.
Desalination, 173(1), 61-67 (2005)
Mandeep Singh Bakshi et al.
Journal of colloid and interface science, 271(1), 227-231 (2004-02-06)
Mixed micelle formation by tetradecyltrimethylammonium (TTAB) and tetradecyltriphenylphosphonium bromides (TTPB) with benzyldimethyltetradecylammonium chloride (BTDACl) was studied with the help of conductivity and Kraft point measurements. The BTDACl + TTAB mixtures showed synergistic interactions whereas those of BTDACl + TTPB indicated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service