01-5190
Ammonium thiocyanate
SAJ special grade, ≥99%
Synonym(s):
Ammonium rhodanide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NH4SCN
CAS Number:
Molecular Weight:
76.12
Beilstein:
3595135
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
Recommended Products
grade
SAJ special grade
vapor pressure
0.000114 hPa ( 20 °C)
Assay
≥99%
form
solid
availability
available only in Japan
pH
4.0-5.5 (25 °C, 76.1 g/L)
mp
152-154 °C (lit.)
SMILES string
N.SC#N
InChI
1S/CHNS.H3N/c2-1-3;/h3H;1H3
InChI key
SOIFLUNRINLCBN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1
Supplementary Hazards
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Xiaolian Gu et al.
The Journal of investigative dermatology, 135(8), 2077-2083 (2015-04-02)
Epigenetic modifications by DNA methylation are associated with a wide range of diseases. Previous studies in psoriasis have concentrated on epigenetic changes in immune cells or in total skin biopsies that include stromal-associated changes. In order to improve our understanding
Fumi Miyagawa et al.
Journal of immunology (Baltimore, Md. : 1950), 181(2), 1109-1119 (2008-07-09)
To elucidate the mechanisms controlling peripheral tolerance, we established two transgenic (Tg) mouse strains expressing different levels of membrane-bound OVA (mOVA) as a skin-associated self-Ag. When we transferred autoreactive TCR-Tg CD8 T cells (OT-I cells), keratin 14 (K14)-mOVA(high) Tg mice
Mark A Zottola
Journal of molecular graphics & modelling, 28(2), 183-186 (2009-07-25)
Cyanide (CN) is considered to be a terrorist chemical weapon due to its ready availability in multi-kilogram quantities and multi-modal means of intoxication. The body uses the sulfur transferase enzyme rhodanese to detoxify cyanide via conversion of cyanide to thiocyanate.
G Almanzar et al.
Journal of immunological methods, 387(1-2), 36-42 (2012-10-02)
Antibody avidity, defined as the strength of binding between antibody and antigen, represents a functional measure of affinity maturation of antibodies. Determination of the antibody avidity is usually performed separating high and low avidity antibodies by dissociating agents, but measurement
Javad Azizian et al.
Molecular diversity, 6(3-4), 223-226 (2004-04-08)
A new one-pot four component procedure for synthesis of densely functionalized pyrroles using commercially available ninhydrin with phosphorane intermediates produced in the reaction between triphenylphosphine, ammonium thiocyanate (or ammonium acetate) and various dialkyl acetylenedicarboxylates was developed.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service