Skip to Content
Merck
Ammonium thiocyanate SAJ special grade, ≥99%

Ammonium thiocyanate

Ammonium thiocyanate SAJ special grade, ≥99%
Synonym(s):
Ammonium rhodanide, Ammonium thiocyanate
Linear Formula:
NH4SCN
CAS Number:
Molecular Weight:
76.12

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade