S4031
Dextran Sulfate Solution
50% (w/v) liquid
Synonym(s):
50% Dextran sulfate solution
Select a Size
Select a Size
About This Item
Recommended Products
Product Name
Dextran Sulfate 50% Solution,
packaging
pkg of 200 mL
Quality Level
manufacturer/tradename
Chemicon®
technique(s)
hybridization: suitable (nucleic acid)
shipped in
ambient
General description
Application
To prepare a solution containing 10% Dextran Sulfate, add 8 ml of hybridization ingredients to 2 ml of 50% Dextran Sulfate Solution.
See the "Hybrisol Nomograph for Dextran Sulfate" link located under the "Protocols" tab at catalog number S4031 product specific page on www.chemicon.com for determining the proper concentration of dextran sulfate needed for various hybridization condition guidelines.
All
Physical form
Storage and Stability
Legal Information
Disclaimer
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service