Skip to Content
Merck
All Photos(2)

Key Documents

MABE967

Sigma-Aldrich

Anti-DNA polymerase delta p125 Antibody, clone 11E10.1

clone 11E10.1, from mouse

Synonym(s):

DNA polymerase delta catalytic subunit, DNA polymerase delta p125, DNA polymerase subunit delta p125

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
11E10.1, monoclonal
application:
WB
species reactivity:
human
technique(s):
western blot: suitable
citations:

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11E10.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... POLD1(5424)

General description

DNA polymerase delta catalytic subunit (EC 2.7.7.7; UniProt P28340; also known as CDC2 homolog, DNA polymerase subunit delta p125) is encoded by the POLD1 (also known as CDC2, MDPL, POLD, CRCS10) gene (Gene ID 5424) in human. DNA replication in eukaryotes involves multiple enzymes including the B family polymerases Pol α, Pol δ, and Pol ɛ. While Pol α initiates DNA synthesis, Pol δ and Pol ɛ replace Pol α after primer extension and perform the bulk of DNA replication. Pol δ and Pol ɛ are heterotetramers in higher eukaryotes comprised of POLD1 and POLE as the respective catalytic subunit, as well as accessory subunits (POLD2/3/4 and POLE2/3/4) that interact with cofactors such as Proliferating Cell Nuclear Antigen (PCNA). Both POLD1 and POLE contain an exonuclease domain (a.a. 268–471 of POLE and 304–517 of POLD1) that proof-reading capability during DNA replication. In addition to DNA synthesis, Pol ɛ and Pol δ are believed to participate in chromosomal DNA repairs, including nucleotide excision repair (NER), ismatch repair (MMR) and repair of double strand breaks (DSBR). Germline exonuclease domain mutations (EDMs) in POLD1 and POLE cause polymerase proofreading-associated polyposis (PPAP). Tumors with EDMs exhibits dramatically increased base substitutions.

Immunogen

GST-tagged recombinant protein corresponding to the N-terminus of human DNA polymerase delta p125.

Application

Anti-DNA polymerase delta p125 Antibody, clone 11E10.1 is an antibody against DNA polymerase delta p125 Antibody for use in Western Blotting.
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected DNA polymerase delta p125 in 10 µg of Ramos cell lysate.

Quality

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected DNA polymerase delta p125 in 10 µg of Raji cell lysate.

Target description

~125 kDa observed.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service