Skip to Content
Merck
All Photos(1)

Key Documents

1.05360

Millipore

L-Leucine

for biochemistry

Synonym(s):

L-Leucine, Leu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO2
CAS Number:
Molecular Weight:
131.17
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-522-0
NACRES:
NA.21

vapor pressure

<1 hPa ( 20 °C)

Quality Level

pH

5.5-6.5 (20 °C, 20 g/L in H2O)

mp

270-340 °C (decomposition)

solubility

18.9 g/L

density

1.17 g/cm3 at 20 °C

bulk density

300‑400 kg/m3

storage temp.

2-30°C

InChI

1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)

InChI key

ROHFNLRQFUQHCH-UHFFFAOYSA-N

Application


  • Novel recombinant aminoacylase from Paraburkholderia monticola capable of N-acyl-amino acid synthesis.: Highlights the synthetic application of L-Leucine in producing N-acyl-amino acids using engineered enzymes, showcasing its industrial and biotechnological potential (Haeger et al., 2024).

Analysis Note

Assay (perchloric acid titration, calculated on dried substance): ≥ 99.0 %
Identity (IR-spectrum): passes test
Appearance: white to almost white, crystalline powder or shiny flakes
Appearance of solution (0.5 mol/l, hydrochloric acid 1 mol/l): clear and colorless
Spec. rotation (α 20/D, 50 g/l, hydrochloric acid 1 mol/l, calc. on dried substance): +12.2 to +13.2
UV-Absorption (260 nm, 0.5 mol/l, 1 cm, hydrochloric acid 1 mol/l): ≤ 0.05
UV-Absorption (280 nm, 0.5 mol/l, 1 cm, hydrochloric acid 1 mol/l): ≤ 0.02
Chloride (Cl): ≤ 0.02 %
Sulfate (SO₄): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.001 %
As (Arsenic): ≤ 0.0005 %
Ca (Calcium): ≤ 0.001 %
Co (Cobalt): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.0005 %
Mg (Magnesium): ≤ 0.0005 %
Na (Sodium): ≤ 0.005 %
Zn (Zinc): ≤ 0.0005 %
NH₄ (Ammonium): ≤ 0.01 %
Isoleucine: ≤ 1.0 %
other foreign amino acids: ≤ 0.3 %
Other ninhydrine positive substances: ≤ 0.1 %
Loss on drying (105 °C; 3 h): ≤ 0.3 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service