Key Documents
Z117137
Stainless steel 304 syringe needle
gauge 22, L 4 in.
About This Item
UNSPSC Code:
42142600
eCl@ss:
32070500
NACRES:
NB.03
description
Standard Hub with 304 Stainless Steel Tubing
packaging
pack of 1 ea
manufacturer/tradename
Cadence Science 4175
L
4 in.
gauge
22
Related Categories
General description
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Choose from one of the most recent versions:
Certificates of Analysis (COA)