Key Documents
Z558575
Sigma-Aldrich
Aldrich® microscale glassware kit
Economy
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Related Categories
General description
Components:
  • Z684139 Adapter, Claisen, 14/10, (1)
  • Z684155 Adapter, Multipurpose, 14/10, (1)
  • Z684244 Condenser, Jacketed Reflux, 14/10, (1)
  • Z684252 Distillation Head, Hickman w/sideport, (1)
  • Z684260 Flask, Round Bottom, 10 mL, 14/10, (1)
  • Z684295 Spin Vane, 3 & 5 mL, (1)
  • Z684341 Tube, Craig, Complete, 2 mL, (1)
  • Z684368 Tube, Drying, 14/10, (1)
  • Z684422 Vial, Conical, 3.0 mL, (1)
  • Z684430 Vial, Conical, 5.0 mL, (1)
  • Z684201 Plastic Kit Box, (1)
High-quality, low cost glassware kits designed to cover a broad range of organic reactions in student labs and chemical research. Kits contain microscale glassware needed to perform experiments described in the textbook Microscale Organic Laboratory. 14/10 ground-glass joints with holed-caps and O-ring seals provide secure, gas-tight connections.
Economy kit - supplied in a compartmentalized PP storage box with hinged lid. Will do 95% of experiments described in the textbook.
Deluxe kit - supplied in a foam-lined plastic carrying case. Covers all experiments in the textbook.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)