SYNA391420
Synthware™ transfer valve adapter
joint: ST/NS 14/20
Synonym(s):
Synthware A391420, transfer adapter with valve
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Recommended Products
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A391420
Related Categories
General description
The 0-4 mm high vacuum valves have two Viton® O-rings on the shaft and an integral PTFE O-ring on the tip.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)