Skip to Content
Merck
All Photos(1)

Documents

731811

Sigma-Aldrich

Poly(ethylene glycol) diglycidyl ether

average MN 2,000, cross-linking reagent amine reactive

Synonym(s):

Polyethylene glycol, Diepoxy PEG, PEG diglycidyl ether, Polyoxyethylene bis(glycidyl ether)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5O2-(C2H4O)n-C3H5O
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

product name

Poly(ethylene glycol) diglycidyl ether, average Mn 2,000

form

liquid

Quality Level

mol wt

average Mn 2,000

reaction suitability

reagent type: cross-linking reagent
reactivity: amine reactive

mp

49-54 °C

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

2-8°C

InChI

1S/C8H14O4/c1(9-3-7-5-11-7)2-10-4-8-6-12-8/h7-8H,1-6H2

InChI key

AOBIOSPNXBMOAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Impact of Poly(ethylene glycol) Diglycidyl Ether on PVC/PEO Blend Properties: This study examines the influence of PEGDE on the tensile properties, morphology, and electrical conductivity of PVC/PEO blends, highlighting its effectiveness as a surface modifier (Supri et al., 2016).
  • Flexible Wearable Sensors Based on CNTs and PEGDGE: This research focuses on developing flexible wearable sensors by reinforcing carbon nanotubes with PEGDGE, analyzing their strain sensitivity and mechanical properties (Del Bosque et al., 2021).
  • Network Polymers via Addition Reactions of Multifunctional-Amine and PEGDGE: The synthesis of network polymers through the reaction of multifunctional-amine and PEGDGE or diacrylate compounds, exploring their application in materials science (Naga et al., 2020).
  • Modification of Alkali Lignin with PEGDGE for Bio-lubricant Applications: This study modifies alkali lignin with PEGDGE to enhance its use as a thickener in bio-lubricant formulations, demonstrating improved dispersion and stability (Cortés-Triviño et al., 2018).
  • Characterization of Injectable Hyaluronic Acid Dermal Fillers Cross-Linked with PEGDGE: Investigates hyaluronic acid dermal fillers cross-linked with PEGDGE, focusing on their physicochemical properties and applications in aesthetic medicine (Zerbinati et al., 2021).

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Highlighting new synthetic modifications of PEG to improve the mechanical properties and degradation of resulting hydrogels in tissue engineering applications.

Highlighting existing and novel fabrication methods for both, solid and hydrogel-based scaffold for tissue engineering applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service