Skip to Content
Merck
All Photos(2)

Documents

710385

Sigma-Aldrich

Bis(3,5-dimethyl-4-methoxyphenyl)chlorophosphine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22ClO2P
CAS Number:
Molecular Weight:
336.79
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.597

density

1.141 g/cm3 at 25 °C

functional group

phosphine

SMILES string

COc1c(C)cc(cc1C)P(Cl)c2cc(C)c(OC)c(C)c2

InChI

1S/C18H22ClO2P/c1-11-7-15(8-12(2)17(11)20-5)22(19)16-9-13(3)18(21-6)14(4)10-16/h7-10H,1-6H3

InChI key

JWCZKGYRAINWJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Asymmetric organocatalytic electrophilic phosphination
  • Asymmetric hydroformulation using Taddol-based chiral phosphine-phosphite ligands
  • Synthesis of ferrocene-based chiral diphosphines
  • Alcoholysis of phosphane-boranes
  • Asymmetric hydrogenations of bifep-type biferrocenes

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service