Skip to Content
Merck
All Photos(3)

Documents

686697

Sigma-Aldrich

Chlorodi(o-tolyl)phosphine

Synonym(s):

Chlorobis(o-tolyl)phosphine, Di(o-methylphenyl)phosphine chloride, Di-o-tolylchlorophosphine, Bis(2-methylphenyl)phosphinous chloride, Chlorobis(2-methylphenyl)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14ClP
CAS Number:
Molecular Weight:
248.69
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

63-67 °C

functional group

phosphine

SMILES string

Cc1ccccc1P(Cl)c2ccccc2C

InChI

1S/C14H14ClP/c1-11-7-3-5-9-13(11)16(15)14-10-6-4-8-12(14)2/h3-10H,1-2H3

InChI key

KAAGXBGJRWFWPT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service