Key Documents
Z743562
Write protect tape
clear, L × W 1.6 in. × 0.63 in. , perforated to fit 0.5 mL and 1.5 mL tubes
About This Item
UNSPSC Code:
41122700
NACRES:
NB.17
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Diversified Biotech WRIT-2000
L × W
1.6 in. × 0.63 in. , perforated to fit 0.5 mL and 1.5 mL tubes
color
clear
General description
A Clear Adhesive Overwrap to Protect Writing
Resistant - Defend writing against organic solvents and caustic agents
Easy Application - Comes in a handy dispenser and is perforated to fit 0.5ml and 1.5ml tubes.
Multi-Surface - Adhere to all plastics, glass and metals
Temperature Range - -196 C to 150 C
Choose from one of the most recent versions:
Certificates of Analysis (COA)