Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200656

Sigma-Aldrich

Anti-Bovine Albumin antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-BSA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~70 kDa

species reactivity

bovine, human

concentration

~1 mg/mL

technique(s)

immunoblotting: 0.03-0.06 μg/mL
immunoprecipitation (IP): 2.5-5 μg
indirect ELISA: 0.25-0.5 μg/mL (using 5μg/mL BSA for coating)

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

bovine ... Alb(280717)
human ... ALB(213)

General description

Serum albumin, the most abundant protein (30-50 mg/mL) in plasma, is a multifunctional non-glycosylated, negatively charged protein produced in the liver. It binds a wide variety of lipophilic compounds such as steroids and lipophilic hormones.

Immunogen

purified BSA from Cohn Fraction V

Application

Anti-Bovine Albumin antibody produced in rabbit has been used in:
  • ELISA
  • immunoblotting
  • immunoprecipitation

Biochem/physiol Actions

Albumin functions include regulation of osmotic pressure in blood and transport of fatty acids and other lipophilic compounds. Serum albumin has been reported as a valuable biomarker in diseases such as cancer, HIV diseases that need monitoring of glycemic control.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samantha Knott et al.
Frontiers in microbiology, 12, 655873-655873 (2021-05-18)
Biofilms are typically studied in bacterial media that allow the study of important properties such as bacterial growth. However, the results obtained in such media cannot take into account the bacterial localization/clustering caused by bacteria-protein interactions in vivo and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service