Key Documents
Z118885
Spinbar® magnetic stirring flea assortment
pack of 12 ea
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
Pricing and availability is not currently available.
material
PTFE-coated
cylindrical PTFE-coated bar
packaging
pack of 12 ea
manufacturer/tradename
Bel-Art F37186-0000
General description
1 of each of blue, red and yellow: 8 × 1.5 mm, 5 × 2 mm, 7 × 2 mm, 10 × 3 mm.
Legal Information
Spinbar is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)