CLS703401L
Pyrex® VISTA™ filter flask, heavy-walled
capacity 1000 mL
Synonym(s):
conical flasks, filtering flask, filtering flasks, flasks erlenmeyer, Corning® 70340
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
flat bottom flask
narrow-mouth
sterility
non-sterile
feature
closure type Rubber stoppers
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 70340-1L
capacity
1000 mL
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)