XX1014705
Millipore Vacuum Filtering Side-Arm Flask
1L, Threaded side-arm
Synonym(s):
Vacuum Flask
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
technique(s)
vacuum filtration: suitable
flask capacity
1 L
fitting
No. 8 outlet stopper for standard 1 L filtering flask (perforated silicone)
application(s)
filtration
shipped in
ambient
General description
Millipore Vacuum Filtering Side-Arm Flask
1L, Threaded side-arm
These side-arm flasks are designed for use in vacuum filtration procedures with classic Millipore Glass Filter Holders.
This item is an accessory item for multiple kits.
Features & Benefits:
• Side arm connects to vacuum source with 3/8 in. I.D. hose
• 1 L and 4 L flasks accept No. 8 perforated stopper
• 125 mL flask accepts No. 5 stopper
Applications:
Vacuum Filtration Using Classic Filter Holders
1L, Threaded side-arm
These side-arm flasks are designed for use in vacuum filtration procedures with classic Millipore Glass Filter Holders.
This item is an accessory item for multiple kits.
Features & Benefits:
• Side arm connects to vacuum source with 3/8 in. I.D. hose
• 1 L and 4 L flasks accept No. 8 perforated stopper
• 125 mL flask accepts No. 5 stopper
Applications:
Vacuum Filtration Using Classic Filter Holders
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service