23987
Economy Moisture Trap
volume 400 cc, 1/8 in. fitting
Synonym(s):
ECONOMY MOISTURE-REMOVING TRAP, 400CC, 1/8IN FITTINGS
About This Item
Recommended Products
volume
400 cc
fitting
1/8 in. fitting
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The indicating 4A changes from blue to tan at 20% relative humidity.
Note: Not for use with carrier gas streams.
Dimensions:
- 400 cc Traps: 17 1/2 in. (44.4 cm) long x 2 in. (51 mm) diameter
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 Inhalation
Target Organs
Lungs
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service