Z740234
TLC Aluminum Sheets
L × W 20 cm × 20 cm, silica gel 60 matrix (C18), binder, Polymeric, fluorescent indicator
Synonym(s):
HPTLC (High Performance TLC) Aluminum Sheets
About This Item
Recommended Products
material
aluminum support
silica gel 60 matrix (C18)
Quality Level
feature
binder Polymeric
fluorescent indicator
packaging
pkg of 20 ea
manufacturer/tradename
EMD Millipore® 1.05559.0001
technique(s)
HPTLC: suitable
thin layer chromatography (TLC): suitable
L × W
20 cm × 20 cm
layer thickness
250 μm
particle size
10-12 μm
pore size
60 Å pore size
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Application of TLC-Spectrodensitometric and Chemometric Methods for Determination of Momenta Cream: A Comparative Study Applied on Ternary Mixture.: The research compares TLC-spectrodensitometric and chemometric methods for simultaneous determination of components in Momenta cream using aluminum TLC plates. The study validates these methods as effective for analyzing pharmaceutical formulations (Salem et al., 2021).
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service