Z740227
TLC Aluminum Sheets
L × W 20 cm × 20 cm, silica gel 60 matrix, binder, Polymeric, fluorescent indicator: no
Synonym(s):
Thin Layer Chromatography Aluminum Sheets
About This Item
Recommended Products
material
aluminum support
silica gel 60 matrix
Quality Level
feature
binder Polymeric
fluorescent indicator: no
packaging
pkg of 25 ea
manufacturer/tradename
EMD Millipore® 1.05553.0001
technique(s)
thin layer chromatography (TLC): suitable
L × W
20 cm × 20 cm
layer thickness
200 μm
particle size
10-12 μm
pore size
60 Å medium pore diameter
Looking for similar products? Visit Product Comparison Guide
Application
- Synthesis and Antimicrobial Evaluation of Some Novel Pyrimidine, Pyrazole, Chromene and Tetrahydrobenzo[b]thiophene Derivatives Bearing Pyrimidinthione Moiety: This research utilizes TLC aluminum sheets in chromatography to synthesize and evaluate antimicrobial properties of novel chemical derivatives. The study highlights the importance of TLC aluminum sheets in facilitating the analysis and synthesis of complex organic compounds in material science and pharmaceutical research (Reheim MAMA, Hafiz ISA, Elian MA, 2020).
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service