Skip to Content
Merck
All Photos(1)

Key Documents

56524

Supelco

Silica gel on TLC Al foils

with fluorescence indicator 254 nm

Synonym(s):

Silica gel coated aluminum TLC sheets

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
NACRES:
NC.07

material

aluminum support
silica gel 60 matrix

Quality Level

feature

binder Polymeric
fluorescent indicator

packaging

pkg of 25 ea

manufacturer/tradename

Sigma-Aldrich

technique(s)

thin layer chromatography (TLC): suitable

L × W

20 cm × 20 cm

layer thickness

200 μm

particle size

8.0-12.0 μm

pore size

0.75 mL/g pore volume
60 Å mean pore size

Looking for similar products? Visit Product Comparison Guide

General description

  • Outstanding wettability (even for 100% aqueous visualisation reagents)
  • Easy and precise cutting (no flaking of silica)
  • Excellent separation efficiency and reproducibility from batch to batch
Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium(III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation resulting in the formation of uniform polymer particles with narrow particle size distribution and high bulk density. Silica gel is widely used as layer material for TLC. Plaster of Paris is generally used as binding agent to hold silica gel firmly to the TLC plates. Silica gel is used as sorbent which is applied to aluminium foils.

Application

Silica gel on TLC Alu foils may be used to perform thin layer chromatography.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph C. Touchstone.
Practice of Thin Layer Chromatography (1992)
A simple method for production of pure silica from rice hull ash
Kalapathy, U., A. Proctor, and J. Shultz.
Bioresource Technology, 73, 257-262 (2000)
Silica gel supported zirconocene dichloride/methylalumoxane catalysts for ethylene polymerization: Effects of heterogenation on activity, polymer microstructure and product morphology.
Janiak, Christoph, and Bernhard Rieger.
Angew. Makromol. Chem., 215, 47-57 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service