Skip to Content
Merck
All Photos(1)

Key Documents

SML2659

Sigma-Aldrich

Spastazoline

≥98% (HPLC)

Synonym(s):

(R)-N-(5-(tert-butyl)-1H-pyrazol-3-yl)-2-(3-isopropylpiperazin-1-yl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30N8
CAS Number:
Molecular Weight:
382.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[nH]1nc(cc1Nc2nc(nc4[nH]ccc42)N3C[C@H](NCC3)C(C)C)C(C)(C)C

InChI

1S/C20H30N8/c1-12(2)14-11-28(9-8-21-14)19-24-17-13(6-7-22-17)18(25-19)23-16-10-15(26-27-16)20(3,4)5/h6-7,10,12,14,21H,8-9,11H2,1-5H3,(H3,22,23,24,25,26,27)/t14-/m0/s1

InChI key

HNOLTAKAPDKZRY-AWEZNQCLSA-N

Biochem/physiol Actions

Spastazoline is a cell penetrant, potent and selective ATP-competitive inhibitor of spastin.
cell penetrant, potent and selective ATP-competitive inhibitor of spastin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rudolf Pisa et al.
Journal of the American Chemical Society, 141(14), 5602-5606 (2019-03-16)
The bump-hole approach is a powerful chemical biology strategy to specifically probe the functions of closely related proteins. However, for many protein families, such as the ATPases associated with diverse cellular activities (AAA), we lack structural data for inhibitor-protein complexes
Tommaso Cupido et al.
Nature chemical biology, 15(5), 444-452 (2019-02-20)
Spastin is a microtubule-severing AAA (ATPases associated with diverse cellular activities) protein needed for cell division and intracellular vesicle transport. Currently, we lack chemical inhibitors to probe spastin function in such dynamic cellular processes. To design a chemical inhibitor of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service