Skip to Content
Merck
All Photos(1)

Key Documents

SML2639

Sigma-Aldrich

BDT001

≥98% (HPLC)

Synonym(s):

N-(5-Chloro-2-hydroxyphenyl)-3-(piperidin-1-ylsulfonyl) benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19ClN2O4S
CAS Number:
Molecular Weight:
394.87
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=CC=C(C=C1NC(C2=CC=CC(S(N3CCCCC3)(=O)=O)=C2)=O)Cl

InChI key

GRZNJWNQXQKQQL-UHFFFAOYSA-N

Biochem/physiol Actions

BDT001 is a selective negative allosteric modulator of Flt-3 receptor tyrosine kinase. It showed minimal binding to 25 diverse RTKs and 49 intracellular kinases. In primary cultures of adult DRG neurons, BDT001 reversed the potentiation of capsaicin-induced TRPV1 activation by FL, with a maximal effect at 1 μM. In a mouse study BDT001 alleviated nerve injury-induced peripheral neuropathic pain but had no effect on nociception in non-injured animals.
Negative allosteric modulator of Flt-3 receptor tyrosine kinase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyril Rivat et al.
Nature communications, 9(1), 1042-1042 (2018-03-14)
Peripheral neuropathic pain (PNP) is a debilitating and intractable chronic disease, for which sensitization of somatosensory neurons present in dorsal root ganglia that project to the dorsal spinal cord is a key physiopathological process. Here, we show that hematopoietic cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service