P0066
p70S6K, active, His tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
S6K1, STK14A RPS6KB1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
recombinant
expressed in baculovirus infected Sf9 cells
Quality Level
product line
PRECISIO® Kinase
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
65-89 nmol/min·mg
mol wt
~76 kDa
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... RPS6KB1(6198)
Biochem/physiol Actions
p70S6K is responsible for the phosphorylation of 40S ribosomal protein S6 and is ubiquitously expressed in human adult tissues.p70S6K is activated by serum stimulation and this activation is inhibited by wortmannin and rapamycin. p70S6k activity undergoes changes in the cell cycle and increases 20-fold in G1 cells released from G0. p70S6K activation requires sequential phosphorylations at proline-directed residues in the putative autoinhibitory pseudosubstrate domain, as well as threonine 389, a site phosphorylated by phosphoinositide-dependent kinase 1 (PDK-1).
Physical form
Supplied in 50mM NaPhosphate pH 7.0, 300 mM NaCl, 150 mM imidazole, 0.1mM PMSF, 0.2 mM DTT, 25% glycerol.
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of biological chemistry, 271(2), 963-971 (1996-01-12)
We show here using synchronized Swiss mouse 3T3 fibroblasts that p70 S6 kinase (p70S6k) and mitogen-activated protein kinases (p42mapk/p44mapk) are not only activated at the G0/G1 boundary, but also in cells progressing from M into G1. p70S6k activity increases 20-fold
Critical reviews in biochemistry and molecular biology, 29(6), 385-413 (1994-01-01)
Activation of cell growth leads to the multiple phosphorylation of 40S ribosomal protein S6. The kinase responsible for controling this event is termed p70s6k/p85s6k. Both isoforms of the kinase are derived from a common gene activated by a complex set
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service