Key Documents
HS120042
Microtube Storage Box
Holds 100 x 1.5-2.0 mL tubes, size 100 wells, natural polypropylene, pk of 5
About This Item
UNSPSC Code:
41123300
NACRES:
NB.12
Pricing and availability is not currently available.
material
natural polypropylene
feature
Holds 100 x 1.5-2.0 mL tubes
packaging
pk of 5
manufacturer/tradename
Heathrow Scientific HS120042
L × W × H
5.6 in. × 6 in. × 2.2 in.
size
100 wells
well diam.
12.3 mm
Features and Benefits
  • for larger storage requirements of 1.5-2.0 mL microtubes or cryogenic vials
  • easy-open hinged lid with snap clasp
  • autoclavable
  • designed for quick visual identification
  • uniform wells with location indentification
  • tube capacity is indicated on the outside of the box
  • translucent vibrant colors with frosted labeling area
  • debossed with a distinctive wave design
Choose from one of the most recent versions:
Certificates of Analysis (COA)