GE80-1129-52
EPH electrode wick for MultiPhor™ II electrophoresis unit
W × L 104 mm × 253 mm, pack of 500 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
Recommended Products
grade
electrophoresis grade
packaging
pack of 500 ea
manufacturer/tradename
Cytiva 80112952
W × L
104 mm × 253 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Used for connecting the buffer solution with the gel.
Legal Information
MultiPhor is a trademark of Cytiva
Storage Class Code
13 - Non Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service