Skip to Content
Merck
All Photos(1)

Documents

GE17-0536-01

Superose® 12 Prep Grade

Cytiva 17-0536-01, pack of 125 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NA.56

packaging

pack of 125 mL

manufacturer/tradename

Cytiva 17-0536-01

matrix

highly cross-linked agarose

particle size

30 μm

cleaning in place

1-14

working range

3-12

General description

Good resolution separations of proteins and other biomolecules according to size, Wide separation range – separations in two molecular weight ranges from 1000 up to 5 million, Excellent reproducibility and durability.

Application

Superose® 6 is designed for high-resolution chromatography. Superose® 6 prep grade permits easy, efficient scale-up to preparative separations. Superose® 12 is for high resolution purification of Proteins from 1 × 103 to 3 × 105 molecular weight, while Superose® 12 prep grade is useful for preparative purification of these higher molecular weight Proteins

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Superose is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Understanding pressure drop in chromatography systems is crucial for optimal functionality across different parts of the system.

Protocols

Superose from Cytiva are Size Exclusion Chromatography media consisting of a composite base matrix of dextran and agarose. This page shows how to perform a separation with a superose column.

Superose from Cytiva are Size Exclusion Chromatography media consisting of a composite base matrix of dextran and agarose. This page shows how to perform a separation with a superose column.

Superose from Cytiva are Size Exclusion Chromatography media consisting of a composite base matrix of dextran and agarose. This page shows how to perform a separation with a superose column.

Superose from Cytiva are Size Exclusion Chromatography media consisting of a composite base matrix of dextran and agarose. This page shows how to perform a separation with a superose column.

See All

Related Content

Separation options for Size Exclusion Chromatography with Superose from Cytiva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service