EP3125000184
Eppendorf® Research® Plus Move It pipette
no. of channels, 4, volume 120-1200 μL, cone distance 9-33 mm, dark green
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
dark green
feature
no. of channels 4
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 3125000184
cone distance
9-33 mm
volume
120-1200 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Adjustable tip spacing pipette
- Multi-channel pipette
- 4-channel
- 120-1200μL
- 9mm cone distance
- Dark green
Application
- Forward pipetting
- Reverse pipetting
- Removal of supernatants
- Sample mixing
- Phase extraction
- Filling of plates, gels, and reaction vessel
Features and Benefits
Air cushioned pipettes with variable volume and adjustable tip spacing
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Research is a registered trademark of Eppendorf AG
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service