Skip to Content
Merck
All Photos(1)

Key Documents

EP3125000176

Eppendorf® Research® Plus Move It pipette

no. of channels, 8, volume 30-300 μL, cone distance 4.5-14 mm, orange

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

material

orange

feature

no. of channels 8

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 3125000176

cone distance

4.5-14 mm

volume

30-300 μL

Looking for similar products? Visit Product Comparison Guide

General description

  • Adjustable tip spacing pipette
  • Multi-channel pipette
  • 8-channel
  • 30-300μL
  • 9mm cone distance
  • Orange

Application

  • Forward pipetting
  • Reverse pipetting
  • Removal of supernatants
  • Sample mixing
  • Phase extraction
  • Filling of plates, gels, and reaction vessel

Features and Benefits

Air cushioned pipettes with variable volume and adjustable tip spacing

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Research is a registered trademark of Eppendorf AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service