Skip to Content
Merck
All Photos(1)

Key Documents

NIST2975

Diesel particulate matter

NIST® SRM® 2975

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of 1 g

manufacturer/tradename

NIST®

application(s)

industrial hygiene

format

matrix material

SMILES string

C

InChI

1S/CH4/h1H4

InChI key

VNWKTOKETHGBQD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM 2975 consists of a bottle with 1g of diesel particulate matter, collected from an industrial diesel-powered forklift.

SRM 2975_cert

SRM 2975_SDS

Application

Diesel particulate matter (Industrial forklift) standard reference material (SRM) is intended for use in evaluating analytical methods used to determine selected polycyclic aromatic hydrocarbons (PAHs) and nitro-substituted PAHs (nitro-PAHs) in diesel particulate matter and similar matrices.

Features and Benefits

  • Accompanied with certificate documenting NIST-certified values of selected polycyclic aromatic hydrocarbons (PAHs) and nitro-substituted PAHs (nitro-PAHs) and non-certified values of total extractable mass, particle-size distribution, and specific surface area. 
  • Certified values are metrologically traceable to and SI-derived units of mass fraction.
  • The NIST certificate is provided with storage, use and handling instructions.

Other Notes

Certified for the analytes listed below.
2-Methylphenanthrene, 1-Methylphenanthrene, Triphenylene, Benzo[j]fluoranthene, Benzo[a]fluoranthene, Dibenz[a,h + a,c]anthracene, Picene, Dibenzo[a,e]pyrene, Dibenzo[b,k]fluoranthene

Reference/Informational value is provided for the following analytes.
9-Nitrophenanthrene, 3-Nitrophenanthrene, 2-Nitrofluoranthene, 3-Nitrofluoranthene, 1-Nitropyrene, 7-Nitrobenz[a]anthracene, 6-Nitrochrysene, Naphthalene, Biphenyl, Fluorene, Phenanthrene, Anthracene, 3-Methylphenanthrene, 9-Methylphenanthrene, 1,2-Dimethylphenanthrene, 1,6-, 1,7-,2,5-, and 2,9-Dimethylphenanthrene, 1,8-Dimethylphenanthrene, 2,6-Dimethylphenanthrene, 2,7-Dimethylphenanthrene, 3,6, Dimethylphenanthrene,Fluoranthene, Pyrene, 1-, 3-, and 7 Methylfluoranthene, 2-Methylpyrene, 4-Methylpyrene, Benzo[c]phenanthrene, Benz[a]anthracene, Chrysene, Benzo[k]fluoranthene, Benzo[e]pyrene, Benzo[a]pyrene, Perylene, Indeno[1,2,3-cd]pyrene Benzo[ghi]perylene, Dibenz[a,j]anthracene, Benzo[b]chrysene, Coronene, 2-Methylnaphthalene, 1-Methylnaphthalene, Acenaphthene, Benzo[ghi]fluoranthene, Benzo[b]fluoranthene, 2-Methylnaphthalene, 1-Methylnaphthalene, Acenaphthene, Benzo[ghi]fluoranthene, Benzo[b]fluoranthene, 1-Nitronaphthalene, 2-Nitronaphthalene, 9-Nitroanthracene, 8-Nitrofluoranthene, 4-Nitropyrene, 6-Nitrobenzo[a]pyrene, 1,3-Dinitropyrene, 1,6-Dinitropyrene, 1,8-Dinitropyrene, 9-Nitroanthracene

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduced toxicity of diuron to the freshwater green alga Pseudokirchneriella subcapitata in the presence of black carbon.
Knauer K, et al.
Aquatic Toxicology (Amsterdam, Netherlands), 83(2), 143?148-143?148 (2007)
Black carbon concentrations in California vehicles and estimation of in-vehicle diesel exhaust particulate matter exposures.
Fruin A S, et al.
Atmospheric Environment, 38(25), 4123-4133 (2004)
Stephanie M Bilinovich et al.
International journal of environmental research and public health, 17(20) (2020-10-15)
Autism spectrum disorder (ASD) is a neurodevelopmental disorder characterized by disruptions in social communication and behavioral flexibility. Both genetic and environmental factors contribute to ASD risk. Epidemiologic studies indicate that roadway vehicle exhaust and in utero exposure to diesel particulate
Yung-Ho Hsu et al.
Free radical biology & medicine, 135, 235-244 (2019-03-18)
Traffic emission is responsible for most small-sized particulate matter (PM) air pollution in urban areas. Several recent studies have indicated that traffic-related PM may aggravate kidney disease. Furthermore, exposure to particulate air pollution may be related to the risk of

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What is the approximate PM diameter of this product?

    1 answer
    1. The PM diameter can be found on page 9 of the corresponding Certificate of Analysis from NIST: https://www-s.nist.gov/srmors/certificates/2975.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service