Skip to Content
Merck
All Photos(1)

Documents

51578

Supelco

Heptadecane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15CH3
CAS Number:
Molecular Weight:
240.47
Beilstein:
1738898
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

8.3 (vs air)

vapor pressure

1 mmHg ( 115 °C)

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.436 (lit.)

bp

302 °C (lit.)

mp

20-22 °C (lit.)
21-23 °C

density

0.777 g/mL at 25 °C (lit.)

application(s)

petroleum

format

neat

SMILES string

CCCCCCCCCCCCCCCCC

InChI

1S/C17H36/c1-3-5-7-9-11-13-15-17-16-14-12-10-8-6-4-2/h3-17H2,1-2H3

InChI key

NDJKXXJCMXVBJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heptadecane belongs to the n-alkane class of petroleum hydrocarbons.

Application

Heptadecane may be used as an analytical reference standard for the determination of heptadecane in:
  • Bio-oils of bocaiuva residues by two-dimensional gas chromatography (GC x GC) coupled to time-of-flight mass spectrometry (TOFMS) equipped with electron impact (EI) ionization.
  • Cyanobacterial culture media by head-space solid-phase microextraction (HS-SPME) combined with GC-MS.
  • Soil samples by ultrasonic extraction and silicone chromatography column purification, followed by analysis using GC coupled to flame ionization detector (FID) and GC-MS operating on selected ion monitoring (SIM) detection mode.
  • Stem ethanolic extracts of Cayratia trifolia by GC-MS.
  • Crude oils by SPME-GC combined with isotope ratio mass spectrometry (IRMS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

300.2 °F - closed cup

Flash Point(C)

149 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of new method for D/H ratio measurements for volatile hydrocarbons of crude oils using solid phase micro-extraction (SPME) coupled to gas chromatography isotope ratio mass spectrometry (GC-IRMS).
Li Z, et al.
Marine and Petroleum Geology, 89(6), 232-241 (2018)
Determination of n-alkanes contamination in soil samples by micro gas chromatography functionalized by multi-walled carbon nanotubes.
Li Y, et al.
Chemosphere, 158, 154-162 (2016)
Efficacy of head space solid-phase microextraction coupled to gas chromatography-mass spectrometry method for determination of the trace extracellular hydrocarbons of cyanobacteria.
Guan W, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1029, 113-120 (2016)
Characterization of bio-oils obtained from pyrolysis of bocaiuva residues.
Cardoso CAL, et al.
Renewable Energy, 91, 21-31 (2016)
Chromatographic and spectrophotometric analysis of bioactive compounds from Cayratia trifolia (L.) stem.
Sowmya S, et al.
International Journal of Pharmacy and Pharmaceutical Sciences, 8(6), 57-64 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service