Skip to Content
Merck
All Photos(2)

Documents

H6703

Sigma-Aldrich

Hexadecane

ReagentPlus®, 99%

Synonym(s):

n-Hexadecane, Cetane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14CH3
CAS Number:
Molecular Weight:
226.44
Beilstein:
1736592
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

7.8 (vs air)

Quality Level

vapor pressure

1 mmHg ( 105.3 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

395 °F

refractive index

n20/D 1.434 (lit.)

bp

287 °C (lit.)

mp

18 °C (lit.)

transition temp

solidification point 17.5-18.5 °C

density

0.773 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCC

InChI

1S/C16H34/c1-3-5-7-9-11-13-15-16-14-12-10-8-6-4-2/h3-16H2,1-2H3

InChI key

DCAYPVUWAIABOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexadecane (n-Hexadecane) is an aliphatic long-chain hydrocarbon. It is a reference fuel with the cetane number 100 and is employed in determining the cetane number of diesel fuels. The mechanism of the oxidation of hexadecane in the gas-phase has been described. The density, speed of sound, adiabatic compressibility and compressed liquid density of hexadecane have been determined. The performance of platinum/zeolite catalysts for the hydroisomerization of n-hexadecane has been assessed. The thermal degradation of n-hexadecane forms n-alkanes and 1-alkenes as key products.

Application

Hexadecane has been used as a phase change material (PCM) in the preparation of hexadecane/xGnP (exfoliated graphite nanoplatelet), a composite PCM loaded with xGnP.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

233.6 °F

Flash Point(C)

112 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kinetics and product distribution of n-hexadecane pyrolysis.
Watanabe M, et al.
AIChE Journal, 46(4), 843-856 (2000)
Density and speed of sound measurements of hexadecane.
Outcalt S, et al.
The Journal of Chemical Thermodynamics, 42(6), 700-706 (2010)
High thermal performance composite PCMs loading xGnP for application to building using radiant floor heating system
Jeon J, et al.
Solar Energy Mat. and Solar Cells, 101, 51-56 (2012)
The gas-phase oxidation of n-hexadecane.
Fournet R, et al.
International Journal of Chemical Kinetics, 33(10), 574-586 (2001)
Comparison of Pt/zeolite catalysts for n-hexadecane hydroisomerization.
Park KC and Ihm SK.
Applied Catalysis A: General, 203(2), 201-209 (2000)

Protocols

Assays that predict passive absorption of orally administered drugs have become increasingly important in the drug discovery process. As previously described by Faller and Kansy such assays provide rapid, low cost and automation friendly methods to measure a compound’s passive permeability.

Assays that predict passive absorption of orally administered drugs have become increasingly important in the drug discovery process. As previously described by Faller and Kansy such assays provide rapid, low cost and automation friendly methods to measure a compound’s passive permeability.

Assays that predict passive absorption of orally administered drugs have become increasingly important in the drug discovery process. As previously described by Faller and Kansy such assays provide rapid, low cost and automation friendly methods to measure a compound’s passive permeability.

Assays that predict passive absorption of orally administered drugs have become increasingly important in the drug discovery process. As previously described by Faller and Kansy such assays provide rapid, low cost and automation friendly methods to measure a compound’s passive permeability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service