LUXCSYS
mPAGE® Lux Casting System
for hand casting mini gels using UV light with a 0.75 mm, 1.0mm or 1.5 mm thickness, includes instrument, casters, plates and combs
Synonym(s):
curing system, gel caster
About This Item
Recommended Products
AC/DC input
100 - 240 V, 1.5 A
packaging
pkg of 1 kit (size_0.75mm)
pkg of 1 kit (size_1.0 mm)
pkg of 1 kit (size_1.5 mm)
manufacturer/tradename
mPAGE®
greener alternative product characteristics
Less Hazardous Chemical Syntheses
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
parameter
15-30 °C ambient temp. limit
20-80% humidity limit (non-condensing)
H
<2000 m , Altitude
L × W × H
41.275 cm × 29.21 cm × 32.385 cm
gel size
8.3 cm × 10.1 cm
gel thickness
0.75 mm
1.0 mm
1.5 mm
well size × volume
0.75 mm × 10 well (25 μL) × 15 well (10 μL)
1.0 mm × 10 well (40 μL) × 15 well (25 μL)
1.5 mm × 10 well (60 μL) × 15 well (36 μL)
compatibility
for use with mPAGE®
greener alternative category
General description
Components
- mPAGE® Lux Curing Station
- mPAGE® Lux Mixing Tube
- mPAGE® Caster (GCR2)
- mPAGE® Comb, selected thickness 10 wells
- mPAGE® Comb, selected thickness 15 wells
- mPAGE® Mini Spacer Plate selected thickness
- mPAGE® Lux Masked Short Plates 10 wells
- mPAGE® Lux Masked Short Plates 15 wells
- Power code (YP12 NA plug, YP22 EU plug, YP61 UK plug, YP18 Japan plug or YP03 China plug)
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.
Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.
Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.
Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.
Related Content
Find protein research tools to prepare, isolate, and analyze proteins. Organized by how to extract, protect, purify, enrich, modify, and quantify proteins.
Find protein research tools to prepare, isolate, and analyze proteins. Organized by how to extract, protect, purify, enrich, modify, and quantify proteins.
Find protein research tools to prepare, isolate, and analyze proteins. Organized by how to extract, protect, purify, enrich, modify, and quantify proteins.
Find protein research tools to prepare, isolate, and analyze proteins. Organized by how to extract, protect, purify, enrich, modify, and quantify proteins.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service