Z694231
Troemner Lab-Frame standard kit with stainless steel rods
small, 24 in. x 24 in.
Synonym(s):
laboratory clamps, laboratory support rods
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
size
S
manufacturer/tradename
Troemner 916197
base size
18 in.
frame W × L
24 in. × 24 in.
Looking for similar products? Visit Product Comparison Guide
General description
Standard Lab-Frame kits are available in five sizes and three lattice rod choices:
A range of lab-frames specifically designed to fit within laboratory fume hood include stainless steel rods. Nickel-plated connectors and lab-frame feet are standard in all kits. Lab-frame bases may be permanently mounted to bench top with screws (included).
- traditional center-less ground aluminum
- non-corroding fiberglass
- tough stainless steel
A range of lab-frames specifically designed to fit within laboratory fume hood include stainless steel rods. Nickel-plated connectors and lab-frame feet are standard in all kits. Lab-frame bases may be permanently mounted to bench top with screws (included).
Vertical mounting, great for glassware set ups in small laboratories where space is limited. Frame measures 24 x 24 inches (610 x 610 mm). Base is 18 inches (457 mm) wide for stability.
Components:
Components:
- 2 in. (51 mm) rods (qty 8)
- 18 in. (457 mm) rods (qty 2)
- 24 in. (610 mm) rods (qty 8)
- S-connectors (qty 18)
- Rod end connectors (qty 4)
- Lab-frame feet (qty 4)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service