Skip to Content
Merck
All Photos(1)

Documents

E13830

Sigma-Aldrich

Ethyl 4-bromocrotonate

75%, technical grade

Synonym(s):

Ethyl trans-4-bromo-2-butenoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH=CHCO2C2H5
CAS Number:
Molecular Weight:
193.04
Beilstein:
1721388
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

75%

form

liquid

refractive index

n20/D 1.493 (lit.)

bp

94-95 °C/12 mmHg (lit.)

density

1.402 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)\C=C\CBr

InChI

1S/C6H9BrO2/c1-2-9-6(8)4-3-5-7/h3-4H,2,5H2,1H3/b4-3+

InChI key

FHGRPBSDPBRTLS-ONEGZZNKSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service