Skip to Content
Merck
All Photos(2)

Documents

16505

Sigma-Aldrich

Methyl trans-4-bromo-2-butenoate

technical, ≥90% (GC)

Synonym(s):

Methyl 4-bromocrotonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH=CHCOOCH3
CAS Number:
Molecular Weight:
179.01
Beilstein:
1745755
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (GC)

contains

silver wool as stabilizer

impurities

~3% methyl cis-4-bromo-2-butenoate

refractive index

n20/D 1.501

bp

70 °C/0.03 mmHg (lit.)

density

1.498 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(=O)\C=C\CBr

InChI

1S/C5H7BrO2/c1-8-5(7)3-2-4-6/h2-3H,4H2,1H3/b3-2+

InChI key

RWIKCBHOVNDESJ-NSCUHMNNSA-N

Application

Methyl trans-4-bromo-2-butenoate (Methyl 4-bromocrotonate) was used in the synthesis of highly substituted benzoxepines or benzopyrans and L-α-amino [4,5-3H]adipic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F

Flash Point(C)

92 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Allylpalladium (IV) Intermediate in the Synthesis of Highly Substituted Benzoxepines and Benzopyrans via Reactions of Stable Pallada (II) cycles with Allyl Bromides.
Guo R, et al.
Organometallics, 26(15), 3784-3783 (2007)
J O'Sullivan et al.
The Biochemical journal, 184(2), 421-426 (1979-11-15)
1. delta-(L-alpha-Amino[4,5-3H]adipyl)-L-cysteinyl-D-[4,4-3H]valine has been synthesized from its constituent amino acids, the L-alpha-amino[4,5-3H]adipic acid being obtained by reduction with 3H2 of methyl 5-acetamido-5,5-diethoxycarbonylpent-2-enoate and subsequent decarboxylation and hydrolysis. 2. In a cell-free system prepared by lysis of protoplasts of Cephalosporium acremonium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service