CLS808250
Pyrex® centrifuge tubes, conical with screw cap
capacity 50 mL, with graduations
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
borosilicate glass
cap (rubber liner)
conical bottom tube
sterility
non-sterile
packaging
case of 12
pack of 12
manufacturer/tradename
Corning 8082-50
availability
not available in EU
O.D. × L
28 mm × 150 mm
cap diam.
24 mm
cap thread
24
capacity
50 mL
color
clear
Looking for similar products? Visit Product Comparison Guide
General description
Cap with glued-in white-rubber liner resists effects of temperature and steam. (Screw thread size for the 15 mL tube is 15-415, and for the 5 mL tube is 24-410)
Formed from borosilicate glass. Stress that glass can withstand is greatly decreased by surface flaws such as scratches, chips, and cracks. Always use with correct sleeve or holder.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service