Skip to Content
Merck
All Photos(1)

Key Documents

50826

Sigma-Aldrich

Trihexyltetradecylphosphonium decanoate

≥95.0% (NMR)

Synonym(s):

Tetradecyltrihexylphosphonium decanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)5]3P[OCO(CH2)8CH3](CH2)13CH3
CAS Number:
Molecular Weight:
655.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
grade:
NMR grade
technique(s):
NMR: suitable
Pricing and availability is not currently available.

grade

NMR grade

Assay

≥95.0% (NMR)

technique(s)

NMR: suitable

functional group

phosphine

SMILES string

CCCCCCCCCC([O-])=O.CCCCCCCCCCCCCC[P+](CCCCCC)(CCCCCC)CCCCCC

InChI

1S/C32H68P.C10H20O2/c1-5-9-13-17-18-19-20-21-22-23-24-28-32-33(29-25-14-10-6-2,30-26-15-11-7-3)31-27-16-12-8-4;1-2-3-4-5-6-7-8-9-10(11)12/h5-32H2,1-4H3;2-9H2,1H3,(H,11,12)/q+1;/p-1

InChI key

HQIPXXNWLGIFAY-UHFFFAOYSA-M

General description

Trihexyltetradecylphosphonium decanoate is an ionic liquid that is used as a convenient and efficient solvent for a wide range of chemical reactions due to its unique properties such as its high thermal stability and low volatility, which make it useful in a variety of organic synthesis applications.

Application

Trihexyltetradecylphosphonium decanoate can be used as a demulsifying agent due to its unique solubility and surface-active properties. It is also used as a catalyst in Henry nitroaldol reactions.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Abdul Sattar et al.
ACS omega, 5(33), 21191-21202 (2020-09-03)
Intermolecular interactions between the constituents of a polymer nanocomposite at the polymer-particle interface strongly affect the segmental mobility of polymer chains, correlated with their glass transition behavior, and are responsible for the improved dynamical viscoelastic properties. In this work, we
Annaly Cruz Sotolongo et al.
Talanta, 210, 120614-120614 (2020-01-29)
A preconcentration method based on a novel hybrid sorption nanomaterial consisting in a 3D graphene-Ni foam functionalized with an ionic liquid (IL) was developed for Hg species determination. The capability of different phosphonium-ionic liquids (PILs) to functionalize the hybrid material
Sarah F R Taylor et al.
Physical chemistry chemical physics : PCCP, 19(22), 14306-14318 (2017-05-26)
This study reports on understanding the formation of bubbles in ionic liquids (ILs), with a view to utilising ILs more efficiently in gas capture processes. In particular, the impact of the IL structure on the bubble sizes obtained has been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service